Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sncaip
Entrez Gene ID
67847
Gene Name
synuclein, alpha interacting protein (synphilin)
Gene Aliases
2810407O15Rik, 4933427B05Rik, BB104594, SYPH1
Chromosome Location
Chr.18: 52767811 - 52915935 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199151.1 NP_001186080.1 8 - 9 1778 75
NM_001199153.1 NP_001186082.1 8 - 9 1683 75
NM_001199154.1 NP_001186083.1 8 - 9 1953 75
NM_026408.4 NP_080684.3 8 - 9 1778 75
GenBank mRNA AF348447.1 - 8 - 9 1706 75
AK013040.1 - 6 - 7 1512 75
AK017012.1 - 5 - 6 713 75
AK044974.1 - 8 - 9 1976 75
AK081942.1 - 8 - 9 1957 75
AK082079.1 - 8 - 9 1782 75
AK132346.1 - 8 - 9 1881 75
AK160111.1 - 8 - 9 1782 75
BC098489.1 - 8 - 9 1682 75
BC130225.1 - 8 - 9 1668 75
BC152544.1 - 8 - 9 1666 75

View Full Product Details