The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032396.3 | NP_115772.2 | 4 - 5 | 553 | 74 |
GenBank mRNA | AB059617.1 | - | 4 - 5 | 516 | 74 |
AK083565.1 | - | 2 - 3 | 161 | 74 | |
AK141321.1 | - | 4 - 5 | 565 | 74 | |
BC049771.1 | - | 4 - 5 | 477 | 74 | |
BC082546.1 | - | 4 - 5 | 553 | 74 | |
BC138464.1 | - | 4 - 5 | 561 | 74 | |
BC138465.1 | - | 4 - 5 | 561 | 74 |