Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wbscr16
Entrez Gene ID
94254
Gene Name
Williams-Beuren syndrome chromosome region 16 homolog (human)
Gene Aliases
5730496C04Rik, AU019812
Chromosome Location
Chr.5: 134148058 - 134177301 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_033572.2 NP_291050.1 7 - 8 1014 76
GenBank mRNA AF410456.1 - 7 - 8 993 76
AK005040.1 - 7 - 8 960 76
AK017733.1 - 7 - 8 1013 76
AK184744.1 - 1 - 2 74 76
AK206545.1 - 1 - 2 76
BC024714.1 - 7 - 8 986 76
BC055335.1 - 7 - 8 1012 76

View Full Product Details