Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Oas3
Entrez Gene ID
246727
Gene Name
2'-5' oligoadenylate synthetase 3
Gene Aliases
Oasl10
Chromosome Location
Chr.5: 120753098 - 120777659 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145226.2 NP_660261.1 15 - 16 3445 79
GenBank mRNA AB067534.1 - 15 - 16 3445 79
AF453830.1 - 15 - 16 3437 79
BC016461.1 - 8 - 9 1543 79
BC141055.1 - 15 - 16 3438 79

View Full Product Details