Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Samd14
Entrez Gene ID
217125
Gene Name
sterile alpha motif domain containing 14
Gene Aliases
AI839049, AI854782, BC034054
Chromosome Location
Chr.11: 95009402 - 95026087 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146025.2 NP_666137.1 4 - 5 1210 112
GenBank mRNA AK044765.1 - 4 - 5 739 112
AK047184.1 - 4 - 5 1187 112
AK049753.1 - 4 - 5 1213 112
BC034054.1 - 4 - 5 734 112
BC049954.1 - 4 - 5 767 112

View Full Product Details