The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153129.2 | NP_694769.2 | 13 - 14 | 1848 | 82 |
GenBank mRNA | AK154563.1 | - | - | 1721 | 82 |
AK164770.1 | - | 14 - 15 | 1834 | 82 | |
AK171969.1 | - | 13 - 14 | 1831 | 82 | |
BC034670.1 | - | 2 - 3 | 214 | 82 | |
BC062886.1 | - | 13 - 14 | 1848 | 82 |