The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153407.2 | NP_700456.1 | 4 - 5 | 991 | 90 |
GenBank mRNA | AB091689.1 | - | 3 - 4 | 740 | 90 |
AK050864.1 | - | 4 - 5 | 1012 | 90 | |
AK146924.1 | - | 4 - 5 | 1011 | 90 | |
AK166557.1 | - | 4 - 5 | 1015 | 90 | |
BC035295.1 | - | 4 - 5 | 991 | 90 |