Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pkn3
Entrez Gene ID
263803
Gene Name
protein kinase N3
Gene Aliases
AW209115, BC034126
Chromosome Location
Chr.2: 30077544 - 30091019 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153805.1 NP_722500.1 13 - 14 1633 59
GenBank mRNA AK142182.1 - 12 - 13 2149 59
AK154801.1 - 12 - 13 2170 59
BC034126.1 - 13 - 14 1633 59

View Full Product Details