The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_170599.2 | NP_733548.2 | 6 - 7 | 1023 | 78 |
GenBank mRNA | AB079880.1 | - | 6 - 7 | 860 | 78 |
AK160146.1 | - | 6 - 7 | 986 | 78 | |
BC057555.1 | - | 6 - 7 | 1023 | 78 |