The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025618.2 | NP_001020789.1 | 1 - 2 | 149 | 52 |
GenBank mRNA | AK017942.1 | - | 1 - 2 | 62 | 52 |
AK049879.1 | - | 1 - 2 | 85 | 52 |