Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Brms1
Entrez Gene ID
107392
Gene Name
breast cancer metastasis-suppressor 1
Gene Aliases
AV003220, AW554636
Chromosome Location
Chr.19: 5041236 - 5049917 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134155.1 NP_598916.1 8 - 9 791 68
GenBank mRNA AF233580.1 - 8 - 9 802 68
AK076729.1 - 8 - 9 799 68
BC016108.1 - 8 - 9 791 68
BC038380.1 - - 3030 68

View Full Product Details