Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Enpep
Entrez Gene ID
13809
Gene Name
glutamyl aminopeptidase
Gene Aliases
6030431M22Rik, APA, Bp-1/6C3, Ly-51, Ly51
Chromosome Location
Chr.3: 129269177 - 129332749 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007934.3 NP_031960.1 3 - 4 1162 76
GenBank mRNA AK031439.1 - 2 - 3 194 76
AK034613.1 - 3 - 4 1134 76
AK128975.1 - 3 - 4 1119 76
AK146808.1 - 3 - 4 1134 76
BC127060.1 - 3 - 4 1026 76
DQ000292.1 - 3 - 4 902 76
M29961.1 - 3 - 4 981 76

View Full Product Details