The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008235.2 | NP_032261.1 | 2 - 3 | 439 | 102 |
GenBank mRNA | AK132536.1 | - | 2 - 3 | 450 | 102 |
AK133538.1 | - | 2 - 3 | 455 | 102 | |
BC018375.1 | - | 2 - 3 | 439 | 102 | |
BC051428.1 | - | 2 - 3 | 440 | 102 |