Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Exoc7
Entrez Gene ID
53413
Gene Name
exocyst complex component 7
Gene Aliases
Exo70, sec70
Chromosome Location
Chr.11: 116287998 - 116306749 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162872.1 NP_001156344.1 10 - 11 1337 66
NM_016857.2 NP_058553.2 11 - 12 1430 66
GenBank mRNA AF014461.1 - 11 - 12 1365 66
AK087601.1 - 10 - 11 1325 66
AK140450.1 - 11 - 12 1404 66
AK162868.1 - 10 - 11 1750 66
AK167769.1 - 10 - 11 1320 66
AK169928.1 - 10 - 11 1324 66
AK173094.1 - 9 - 10 5917 66
BC028927.1 - 10 - 11 1313 66
BC053710.1 - 10 - 11 1324 66

View Full Product Details