The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021503.2 | NP_067478.1 | 1 - 2 | 167 | 107 |
GenBank mRNA | AJ252148.1 | - | 1 - 2 | 129 | 107 |
AK003634.1 | - | 1 - 2 | 107 | ||
AK085927.1 | - | 1 - 2 | 132 | 107 | |
AY013296.1 | - | 1 - 2 | 107 | ||
BC024360.1 | - | 1 - 2 | 129 | 107 |