The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021503.2 | NP_067478.1 | 4 - 5 | 558 | 115 |
GenBank mRNA | AJ252148.1 | - | 4 - 5 | 520 | 115 |
AK003634.1 | - | 4 - 5 | 508 | 115 | |
AY013296.1 | - | 4 - 5 | 534 | 115 | |
BC024360.1 | - | 4 - 5 | 520 | 115 |