Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crot
Entrez Gene ID
74114
Gene Name
carnitine O-octanoyltransferase
Gene Aliases
1200003H03Rik
Chromosome Location
Chr.5: 8966023 - 8997146 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023733.3 NP_076222.1 17 - 18 1880 95
GenBank mRNA AK004567.2 - 17 - 18 1881 95
BC006593.1 - 17 - 18 1868 95
BC012308.1 - 17 - 18 1895 95

View Full Product Details