The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025573.3 | NP_079849.1 | 1 - 2 | 327 | 81 |
NR_036616.1 | 1 - 2 | 327 | 81 | ||
GenBank mRNA | AK011621.1 | - | 1 - 2 | 287 | 81 |
AK020084.1 | - | 1 - 2 | 99 | 81 | |
AK159190.1 | - | 1 - 2 | 325 | 81 | |
BC012217.1 | - | 1 - 2 | 327 | 81 | |
CJ162734.1 | - | 1 - 2 | 310 | 81 |