Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abhd5
Entrez Gene ID
67469
Gene Name
abhydrolase domain containing 5
Gene Aliases
1300003D03Rik, 2010002J10Rik, CDS, CGI-58, IECN5, NCIE2
Chromosome Location
Chr.9: 122351539 - 122381524 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026179.2 NP_080455.1 6 - 7 1135 74
GenBank mRNA AK004873.1 - 6 - 7 1021 74
AK007421.1 - 6 - 7 1017 74
AK019488.2 - 4 - 5 561 74
AK028453.1 - 6 - 7 1021 74
AK050377.1 - 6 - 7 1020 74
AK152717.1 - 7 - 8 1069 74
BC006683.1 - 3 - 4 361 74
BC037063.1 - 6 - 7 1017 74

View Full Product Details