Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nop9
Entrez Gene ID
67842
Gene Name
NOP9 nucleolar protein
Gene Aliases
2610027L16Rik
Chromosome Location
Chr.14: 55745693 - 55755634 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026403.3 NP_080679.3 1 - 2 327 89
GenBank mRNA AK011570.1 - 1 - 2 317 89
AK015585.1 - 1 - 2 77 89
AK032873.1 - 1 - 2 311 89
AK037831.1 - 1 - 2 315 89
AK050912.1 - 1 - 2 317 89
AK083861.1 - 1 - 2 196 89
AK139689.1 - 1 - 2 277 89
AK146020.1 - 1 - 2 282 89
AK146687.1 - 1 - 2 298 89
AK152732.1 - 1 - 2 301 89
AK154387.1 - 1 - 2 319 89
AK159798.1 - 1 - 2 301 89
BC018523.1 - 1 - 2 307 89

View Full Product Details