Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nop9
Entrez Gene ID
67842
Gene Name
NOP9 nucleolar protein
Gene Aliases
2610027L16Rik
Chromosome Location
Chr.14: 55745693 - 55755634 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026403.3 NP_080679.3 4 - 5 1032 80
GenBank mRNA AK011570.1 - 4 - 5 1022 80
AK015585.1 - 4 - 5 782 80
AK032873.1 - 4 - 5 1016 80
AK037831.1 - 4 - 5 1020 80
AK050912.1 - 4 - 5 1023 80
AK083861.1 - 4 - 5 901 80
AK139689.1 - 4 - 5 983 80
AK146020.1 - 4 - 5 987 80
AK146687.1 - 4 - 5 1004 80
AK152732.1 - 4 - 5 1006 80
AK154387.1 - 4 - 5 1024 80
AK159798.1 - 4 - 5 1006 80
AK178392.1 - 1 - 2 152 80
BC018523.1 - 4 - 5 1012 80

View Full Product Details