The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026441.4 | NP_080717.2 | 4 - 5 | 672 | 60 |
GenBank mRNA | AK007394.1 | - | 4 - 5 | 614 | 60 |
AK013238.1 | - | 4 - 5 | 614 | 60 | |
AK046247.1 | - | 4 - 5 | 614 | 60 | |
AK075978.1 | - | 4 - 5 | 677 | 60 | |
BC019191.1 | - | 4 - 5 | 620 | 60 |