Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Chid1
Entrez Gene ID
68038
Gene Name
chitinase domain containing 1
Gene Aliases
3110023E09Rik, AI841869
Chromosome Location
Chr.7: 141493136 - 141539888 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142681.1 NP_001136153.1 3 - 4 392 85
NM_026522.5 NP_080798.2 3 - 4 392 85
GenBank mRNA AK014071.1 - 3 - 4 341 85
AK014141.1 - 3 - 4 367 85
AK036717.1 - 3 - 4 336 85
AK050655.1 - 3 - 4 340 85
AK147377.1 - 3 - 4 355 85
AK160156.1 - 3 - 4 352 85
AK160200.1 - 3 - 4 352 85
BC006876.1 - 3 - 4 341 85
BC061063.1 - 3 - 4 352 85

View Full Product Details