Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Zmym1
- Entrez Gene ID
-
68310
- Gene Name
- zinc finger, MYM domain containing 1
- Gene Aliases
-
5830412B09Rik, AW209050
- Chromosome Location
-
Chr.4: 127047094 - 127061171 on Build GRCm38
- Gene Symbol
- Sfpq
- Entrez Gene ID
-
71514
- Gene Name
- splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated)
- Gene Aliases
-
1110004P21Rik, 2810416M14Rik, 5730453G22Rik, 9030402K04Rik, AU021830, D4Ertd314e, Gm12940, OTTMUSG00000009329, PSF, REP1
- Chromosome Location
-
Chr.4: 127020560 - 127037016 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK031234.1 |
- |
- |
|
75 |
View Full Product Details