The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028523.3 | NP_082799.2 | 14 - 15 | 1922 | 80 |
GenBank mRNA | AF387548.1 | - | 14 - 15 | 2014 | 80 |
AK006805.1 | - | 4 - 5 | 625 | 80 | |
AK051889.1 | - | 12 - 13 | 1200 | 80 | |
AK084652.1 | - | 3 - 4 | 753 | 80 | |
AK165117.1 | - | 2 - 3 | 854 | 80 | |
BC066097.1 | - | 14 - 15 | 1916 | 80 |