Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ide
Entrez Gene ID
15925
Gene Name
insulin degrading enzyme
Gene Aliases
1300012G03Rik, 4833415K22Rik, AA675336, AI507533
Chromosome Location
Chr.19: 37268741 - 37334544 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031156.3 NP_112419.3 22 - 23 2922 84
GenBank mRNA AJ278422.1 - 22 - 23 2942 84
AK004972.2 - 22 - 23 2924 84
AK128986.1 - 22 - 23 2939 84
BC041675.1 - 22 - 23 2954 84

View Full Product Details