The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053147.3 | NP_444377.3 | 1 - 1 | 275 | 58 |
GenBank mRNA | AF326315.1 | - | 1 - 1 | 147 | 58 |
AK043492.1 | - | 1 - 1 | 276 | 58 | |
AK143176.1 | - | 1 - 1 | 756 | 58 | |
AY013784.1 | - | 1 - 1 | 147 | 58 |