The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053246.3 | NP_444476.1 | 8 - 9 | 1182 | 100 |
GenBank mRNA | AF418207.1 | - | 7 - 8 | 1066 | 100 |
AK032352.1 | - | 8 - 9 | 1172 | 100 | |
AK043670.1 | - | 7 - 8 | 1094 | 100 | |
AK077612.1 | - | 8 - 9 | 1174 | 100 | |
AK139363.1 | - | 7 - 8 | 1071 | 100 | |
AK142621.1 | - | 7 - 8 | 1089 | 100 | |
AK156485.1 | - | 8 - 9 | 1182 | 100 | |
BC004705.1 | - | 8 - 9 | 1143 | 100 |