Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Foxp3
Entrez Gene ID
20371
Gene Name
forkhead box P3
Gene Aliases
JM2, scurfin, sf
Chromosome Location
Chr.X: 7579676 - 7595243 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199347.1 NP_001186276.1 5 - 6 653 55
NM_001199348.1 NP_001186277.1 4 - 5 511 55
NM_054039.2 NP_473380.1 4 - 5 586 55
GenBank mRNA AF277991.1 - 4 - 5 511 55
AF277992.1 - 4 - 5 569 55
AK157447.1 - 5 - 6 654 55
AY357712.1 - 3 - 4 369 55
AY357713.1 - 3 - 4 369 55
BC132333.1 - 3 - 4 357 55
BC132335.1 - 3 - 4 357 55
DQ387959.1 - 5 - 6 653 55
KF471319.1 - 3 - 4 353 55

View Full Product Details