The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008786.2 | NP_032812.2 | 1 - 2 | 512 | 79 |
GenBank mRNA | AK006162.1 | - | 1 - 2 | 190 | 79 |
AK148789.1 | - | 1 - 2 | 537 | 79 | |
BC040750.1 | - | 1 - 2 | 105 | 79 | |
BC049613.1 | - | 1 - 2 | 216 | 79 | |
BC058966.1 | - | 1 - 2 | 201 | 79 | |
BY099821.1 | - | 1 - 2 | 244 | 79 | |
D38023.1 | - | 1 - 2 | 176 | 79 | |
M60320.1 | - | 1 - 2 | 176 | 79 |