The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110300.1 | NP_001103770.1 | 7 - 8 | 986 | 59 |
NM_009678.2 | NP_033808.2 | 7 - 8 | 980 | 59 | |
GenBank mRNA | AF067146.1 | - | 7 - 8 | 954 | 59 |
AK010478.1 | - | 7 - 8 | 965 | 59 | |
AK082674.1 | - | - | 843 | 59 | |
BC003704.1 | - | 7 - 8 | 919 | 59 |