Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pygm
Entrez Gene ID
19309
Gene Name
muscle glycogen phosphorylase
Gene Aliases
AI115133, PG
Chromosome Location
Chr.19: 6384429 - 6398459 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011224.1 NP_035354.1 13 - 14 1670 72
GenBank mRNA AF124787.1 - 13 - 14 1670 72
BC012961.1 - 13 - 14 1676 72

View Full Product Details