Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Bop1
Entrez Gene ID
12181
Gene Name
block of proliferation 1
Gene Aliases
AU020183, AW146150, D18861, Erb1p, Kiaa0124, mKIAA0124
Chromosome Location
Chr.15: 76452996 - 76477269 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013481.1 NP_038509.1 11 - 12 1435 69
GenBank mRNA AK079627.1 - 11 - 12 1415 69
AK129058.1 - 11 - 12 1408 69
AK144095.1 - 11 - 12 1446 69
AK167110.1 - 11 - 12 1418 69
AK167225.1 - 11 - 12 1419 69
AK167829.1 - 11 - 12 1413 69
AK167878.1 - 11 - 12 1423 69
AK168127.1 - 11 - 12 1416 69
AK190367.1 - 1 - 2 58 69
AK191522.1 - 1 - 2 58 69
BC012693.1 - 11 - 12 1409 69
U77415.1 - 11 - 12 1435 69

View Full Product Details