Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Actl6a
Entrez Gene ID
56456
Gene Name
actin-like 6A
Gene Aliases
2810432C06Rik, AI851094, ARP4, Actl6, Baf53a, C79802
Chromosome Location
Chr.3: 32708546 - 32726971 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019673.2 NP_062647.2 12 - 13 1431 103
GenBank mRNA AF041476.1 - 12 - 13 1251 103
AK002724.1 - 12 - 13 1226 103
AK090154.1 - 12 - 13 1250 103
AK167905.1 - 12 - 13 1245 103
AK169278.1 - 12 - 13 1250 103
BC001994.1 - 12 - 13 1219 103
BC094498.1 - 12 - 13 1247 103

View Full Product Details