The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020330.4 | NP_065063.1 | 2 - 2 | 1864 | 61 |
GenBank mRNA | AF251559.1 | - | 1 - 1 | 1356 | 61 |
AK014827.2 | - | 2 - 2 | 1865 | 61 | |
BC132152.1 | - | 1 - 1 | 1407 | 61 | |
BC132344.1 | - | 1 - 1 | 1407 | 61 |