The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109761.2 | NP_001103231.1 | 4 - 5 | 646 | 84 |
NM_001177799.1 | NP_001171270.1 | 4 - 5 | 913 | 84 | |
NM_007601.3 | NP_031627.2 | 4 - 5 | 913 | 84 | |
GenBank mRNA | AF091998.1 | - | 4 - 5 | 610 | 84 |
AF127766.2 | - | 4 - 5 | 633 | 84 | |
BC090661.1 | - | 4 - 5 | 646 | 84 | |
BC139790.1 | - | 4 - 5 | 633 | 84 | |
X92523.1 | - | 4 - 5 | 633 | 84 |