Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Anapc1
Entrez Gene ID
17222
Gene Name
anaphase promoting complex subunit 1
Gene Aliases
2610021O03Rik, AI047775, AI853536, AW547281, Apc1, Mcpr, tsg24
Chromosome Location
Chr.2: 128610083 - 128687410 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008569.2 NP_032595.2 2 - 3 483 80
GenBank mRNA AK052404.1 - 2 - 3 464 80
AK053383.1 - 1 - 2 1556 80
AK054305.1 - 2 - 3 465 80
AK077847.1 - 2 - 3 484 80
AK090134.1 - 2 - 3 466 80
AK149762.1 - 2 - 3 455 80
AK164804.1 - 2 - 3 469 80
BC139002.1 - 2 - 3 464 80
BC145492.1 - 2 - 3 464 80
X80169.1 - 2 - 3 468 80

View Full Product Details