The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290457.1 | NP_001277386.1 | 1 - 2 | 251 | 58 |
NM_009046.2 | NP_033072.2 | 1 - 2 | 251 | 58 | |
GenBank mRNA | AK146932.1 | - | 1 - 2 | 252 | 58 |
AK156767.1 | - | 1 - 2 | 207 | 58 | |
BC019765.1 | - | 1 - 2 | 251 | 58 | |
BC034523.1 | - | 1 - 2 | 64 | 58 | |
BC117792.2 | - | 11 - 12 | 58 | ||
BC117793.1 | - | 1 - 2 | 187 | 58 | |
DQ020178.1 | - | 1 - 2 | 61 | 58 | |
DQ020179.1 | - | 1 - 2 | 61 | 58 | |
DQ020180.1 | - | 1 - 2 | 61 | 58 | |
DQ020181.1 | - | 1 - 2 | 61 | 58 | |
M83380.1 | - | 1 - 2 | 88 | 58 | |
S56076.1 | - | 1 - 2 | 226 | 58 |