The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009171.2 | NP_033197.2 | 1 - 2 | 362 | 86 |
GenBank mRNA | AF237702.1 | - | 1 - 2 | 188 | 86 |
AK010439.1 | - | 1 - 2 | 258 | 86 | |
AK132342.1 | - | - | 232 | 86 | |
BC026055.1 | - | - | 227 | 86 | |
BQ942592.1 | - | 1 - 2 | 92 | 86 | |
CF159149.1 | - | 1 - 2 | 362 | 86 | |
X94478.1 | - | 1 - 2 | 188 | 86 | |
X94479.1 | - | 1 - 2 | 188 | 86 |