Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Casq1
Entrez Gene ID
12372
Gene Name
calsequestrin 1
Gene Aliases
CSQ, CSQ-1, CSQ1, sCSQ
Chromosome Location
Chr.1: 172209894 - 172219895 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC064057.1 - 8 - 9 1026 64
U93291.1 - 8 - 9 1044 64

View Full Product Details