The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109873.1 | NP_001103343.1 | 9 - 10 | 1571 | 75 |
NM_009824.2 | NP_033954.2 | 9 - 10 | 1552 | 75 | |
NM_177289.1 | NP_796263.1 | 8 - 9 | 1493 | 75 | |
GenBank mRNA | AF038029.1 | - | 9 - 10 | 1434 | 75 |
AK033680.1 | - | 8 - 9 | 1389 | 75 | |
AK041887.1 | - | 9 - 10 | 1574 | 75 | |
BC055951.1 | - | 1 - 2 | 136 | 75 | |
BC141396.1 | - | 9 - 10 | 1381 | 75 | |
BC151156.1 | - | 9 - 10 | 1381 | 75 |