The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110228.1 | NP_001103698.1 | 3 - 4 | 430 | 92 |
NM_001110229.1 | NP_001103699.1 | 4 - 5 | 517 | 92 | |
NM_001110230.1 | NP_001103700.1 | 4 - 5 | 517 | 92 | |
NM_001110231.1 | NP_001103701.1 | 6 - 7 | 492 | 92 | |
NM_001110232.1 | NP_001103702.1 | 5 - 6 | 503 | 92 | |
NM_001160292.1 | NP_001153764.1 | 4 - 5 | 517 | 92 | |
NM_001160293.1 | NP_001153765.1 | 3 - 4 | 430 | 92 | |
NM_010160.2 | NP_034290.2 | 3 - 4 | 430 | 92 | |
GenBank mRNA | AF090696.1 | - | 5 - 6 | 503 | 92 |
AF090697.1 | - | 3 - 4 | 403 | 92 | |
AK135166.1 | - | 3 - 4 | 381 | 92 | |
AK137292.1 | - | 4 - 5 | 458 | 92 | |
AK151818.1 | - | 2 - 3 | 288 | 92 | |
AK152628.1 | - | 2 - 3 | 288 | 92 | |
AK153267.1 | - | 2 - 3 | 288 | 92 | |
AK160861.1 | - | 6 - 7 | 493 | 92 | |
AK164914.1 | - | 4 - 5 | 458 | 92 | |
AK172648.1 | - | 2 - 3 | 288 | 92 | |
BC026856.1 | - | 3 - 4 | 381 | 92 | |
CF536042.1 | - | 3 - 4 | 430 | 92 | |
Y18298.1 | - | 2 - 3 | 296 | 92 |