The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288586.1 | NP_001275515.1 | 8 - 9 | 868 | 113 |
NM_010786.4 | NP_034916.1 | 9 - 10 | 953 | 113 | |
GenBank mRNA | AK004719.1 | - | 9 - 10 | 912 | 113 |
AK004881.1 | - | 7 - 8 | 2095 | 113 | |
AK088638.1 | - | 9 - 10 | 883 | 113 | |
AK150530.1 | - | 7 - 8 | 644 | 113 | |
AK152685.1 | - | 7 - 8 | 644 | 113 | |
AK160069.1 | - | 8 - 9 | 744 | 113 | |
AK168690.1 | - | 9 - 10 | 936 | 113 | |
BC050902.1 | - | 9 - 10 | 869 | 113 | |
BC092270.1 | - | 9 - 10 | 900 | 107 | |
U47934.1 | - | 7 - 8 | 659 | 113 | |
X58876.1 | - | 9 - 10 | 860 | 113 |