The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166450.2 | NP_001159922.1 | 7 - 8 | 918 | 81 |
NM_001290703.1 | NP_001277632.1 | 9 - 10 | 1107 | 81 | |
NM_001290704.1 | NP_001277633.1 | 9 - 10 | 1098 | 81 | |
NM_001290705.1 | NP_001277634.1 | 6 - 7 | 673 | 81 | |
NM_011398.3 | NP_035528.1 | 7 - 8 | 1005 | 81 | |
GenBank mRNA | AF076981.1 | - | 7 - 8 | 1006 | 81 |
AF155812.1 | - | 6 - 7 | 593 | 81 | |
AF155813.1 | - | 6 - 7 | 584 | 81 | |
AF535150.1 | - | 9 - 10 | 1099 | 81 | |
AF535151.1 | - | 9 - 10 | 1108 | 81 | |
AK148277.1 | - | 7 - 8 | 2453 | 81 | |
BC048692.1 | - | 6 - 7 | 728 | 81 |