The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011452.2 | NP_035582.1 | 1 - 2 | 185 | 150 |
GenBank mRNA | AK005491.1 | - | 1 - 2 | 186 | 150 |
U96705.2 | - | 1 - 2 | 186 | 150 |