Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sycp3
Entrez Gene ID
20962
Gene Name
synaptonemal complex protein 3
Gene Aliases
Cor1, Scp3
Chromosome Location
Chr.10: 88459587 - 88473236 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011517.2 NP_035647.2 2 - 3 262 134
GenBank mRNA AK131949.1 - 2 - 3 186 134
BC048483.1 - 2 - 3 185 134
BC089540.1 - 2 - 3 185 134
BC099552.1 - 2 - 3 185 134
BC132079.1 - 2 - 3 263 134
BC138509.1 - 2 - 3 263 134
Y08485.1 - 2 - 3 271 134

View Full Product Details