The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016762.2 | NP_058042.2 | 10 - 11 | 1827 | 81 |
GenBank mRNA | AK083151.1 | - | 10 - 11 | 1824 | 81 |
AK138211.1 | - | 10 - 11 | 1824 | 81 | |
AK163818.1 | - | 10 - 11 | 1786 | 81 | |
BC005429.1 | - | 10 - 11 | 1697 | 81 | |
BC092298.1 | - | 10 - 11 | 1770 | 81 | |
U69262.1 | - | 10 - 11 | 1826 | 81 |