The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019579.3 | NP_062525.1 | 14 - 15 | 2261 | 72 |
GenBank mRNA | AF199008.1 | - | 14 - 15 | 2192 | 72 |
AK133187.1 | - | 14 - 15 | 2154 | 72 | |
BC067404.1 | - | 14 - 15 | 2122 | 72 | |
BC075698.1 | - | 14 - 15 | 2214 | 72 | |
BC138624.1 | - | 14 - 15 | 2140 | 72 |