The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168505.1 | NP_001161977.1 | 8 - 9 | 1864 | 111 |
NM_019658.6 | NP_062632.2 | 8 - 9 | 1905 | 111 | |
GenBank mRNA | AF068921.1 | - | 7 - 8 | 1540 | 111 |
AK146447.1 | - | 8 - 9 | 1902 | 111 | |
BC013722.1 | - | 4 - 5 | 440 | 111 | |
BC049775.1 | - | 7 - 8 | 1770 | 111 | |
BC083060.1 | - | 8 - 9 | 1864 | 111 |