Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Clec4e
Entrez Gene ID
56619
Gene Name
C-type lectin domain family 4, member e
Gene Aliases
C86253, Clecsf9, Mincle
Chromosome Location
Chr.6: 123281789 - 123289871 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019948.2 NP_064332.1 3 - 4 342 142
GenBank mRNA AB024717.1 - 3 - 4 342 142
AK036493.1 - 2 - 3 2718 142
AK150528.1 - 3 - 4 342 142
AK150899.1 - 3 - 4 342 142
AK151101.1 - 3 - 4 341 142
AK151912.1 - 3 - 4 341 142
AK153008.1 - 3 - 4 341 142
AK153308.1 - 3 - 4 342 142
AK156437.1 - 3 - 4 342 142
AK170698.1 - 3 - 4 293 142
AK171103.1 - 3 - 4 342 142
BC003218.1 - 3 - 4 300 142
CT010223.1 - 3 - 4 219 142

View Full Product Details